Skip to Content
Merck
All Photos(1)

Key Documents

8.21745

Sigma-Aldrich

trans-Decahydronaphthalene

for synthesis

Synonym(s):

trans-Decahydronaphthalene, trans-Decalin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18
CAS Number:
Molecular Weight:
138.25
MDL number:
UNSPSC Code:
12352001
EC Index Number:
207-771-4

vapor pressure

4.2 hPa ( 20 °C)

Assay

≥96.5% (GC)

form

liquid

autoignition temp.

255 °C

expl. lim.

0.7-4.9 % (v/v)

bp

185 °C/1008 hPa

mp

-32.5--31.5 °C

transition temp

flash point 54 °C

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C10H18/c1-2-6-10-8-4-3-7-9(10)5-1/h9-10H,1-8H2

InChI key

NNBZCPXTIHJBJL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.869 - 0.871
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1C

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service