Skip to Content
Merck
All Photos(1)

Key Documents

8.03010

Sigma-Aldrich

Diethylamine

for synthesis

Synonym(s):

Diethylamine, N-Ethylethanamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€22.40
500 ML
€32.80
1 L
€35.70
2.5 L
€68.80
50 L
€1,050.00

€22.40


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€22.40
500 ML
€32.80
1 L
€35.70
2.5 L
€68.80
50 L
€1,050.00

About This Item

Linear Formula:
(C2H5)2NH
CAS Number:
Molecular Weight:
73.14
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-716-3
NACRES:
NA.22

€22.40


Estimated to ship on16 April 2025


Request a Bulk Order

vapor pressure

253 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

290 °C

potency

540 mg/kg LD50, oral (Rat)
582 mg/kg LD50, skin (Rabbit)

expl. lim.

2.0-11.8 % (v/v)

pH

13 (20 °C, 100 g/L in H2O)

bp

56 °C/1013 mbar

mp

-50 °C

transition temp

flash point -23 °C

density

0.71 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CC)CC

InChI

1S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3

InChI key

HPNMFZURTQLUMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Chemiresistive Sensor Development: Diethylamine was used to enhance the sensitivity of a chemiresistive sensor for CO2 gas monitoring, demonstrating its utility in environmental and industrial applications (Rath et al., 2024).

  • Supercritical Fluid Chromatography: Aromatic stationary phases were synthesized using diethylamine, evaluated based on the linear solvation energy relationship model, which expanded its application in supercritical fluid chromatography (Ge et al., 2024).

  • Nanoparticle Synthesis: Diethylamine facilitated the sol-gel method modifications for synthesizing silver-silica nanoparticles, enhancing their theranostic potential in cancer treatment (Nikolopoulou et al., 2023).

  • Pharmaceutical Analysis: Diethylamine was key in developing a novel enantioselective high-performance liquid chromatography-mass spectrometry method, applied to differentiate dextro- and levo-methorphan in post-mortem samples, showcasing its critical role in forensic and clinical toxicology (Jorbenadze et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.703 - 0.705
Water (K. F.): ≤ 0.10 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service