Skip to Content
Merck

Skip To

8.21024

di-Phosphorus pentasulfide

for synthesis

Synonym(s):

Phosphorus pentasulfide, Diphosphorus pentasulfide, Phosphorus(V) sulfide, Phosphorus(V) sulfide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 g
Check Cart for Availability
€ 49,10
1 kg
Check Cart for Availability
€ 96,30
2.5 kg
Check Cart for Availability
€ 190,00

About This Item

Empirical Formula (Hill Notation):
P2S5
CAS Number:
Molecular Weight:
222.27
NACRES:
NA.22
EC Index Number:
215-242-4
UNSPSC Code:
12352303
MDL number:
Grade:
synthesis grade
Bp:
515 °C

€ 49,10


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

form

powder

autoignition temp.

142 °C

potency

389 mg/kg LD50, oral (Rat), 3160 mg/kg LD50, skin (Rabbit)

concentration

27.8-28.8%

dilution

(for synthesis)

pH

1 (20 °C, 10 g/L in H2O)

bp

515 °C

mp

280-284 °C (lit.)

density

2.09 g/mL at 25 °C (lit.)

bulk density

900‑1100 kg/m3

storage temp.

2-30°C

SMILES string

S=P(=S)SP(=S)=S

InChI

1S/P2S5/c3-1(4)7-2(5)6

InChI key

HWVAJUNEPOKXLF-UHFFFAOYSA-N

Analysis Note

Assay P: 27.8 - 28.8 %

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
2321068.210268.22339
grade

synthesis grade

grade

-

grade

-

grade

synthesis grade

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

crystals

form

liquid

autoignition temp.

142 °C

autoignition temp.

-

autoignition temp.

-

autoignition temp.

-

pH

1 (20 °C, 10 g/L in H2O)

pH

-

pH

-

pH

1.0 (25 °C, 5 g/L in H2O)

bp

515 °C

bp

514 °C

bp

192 °C/1013 hPa (decomposes)

bp

105.3 °C/1013 hPa


signalword

Danger

supp_hazards

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Flam. Sol. 1 - Water-react. 1

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
821024100004022536471945
821024010004022536471938
821024250004022536471952

Questions

  1. What is the difference between this item and Phosphorus pentasulfide (232106)?

    1 answer
    1. These products both fall under the chemical abstract service number 1314-80-3 which means chemically they are the same. The only major difference between the two products is that product 821024 has a purity assay range between 27.8-28.8 % while product 232106 has an purity assay range of 98.5-101.5 %. Both products have different testing methods as well which can be viewed via the product specification sheet:

      https://www.sigmaaldrich.com/specification-sheet/SPEC/MDA/CHEM/8.21024/8210242500
      https://www.sigmaaldrich.com/specification-sheets/754/815/232106-BULK_______SIGALD_____.pdf

      Helpful?

Reviews

No rating value

Active Filters