Skip to Content
Merck

Skip To

8.20554

Allyl acetate

for synthesis

Synonym(s):

Allyl acetate, Acetic acid allyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Check Cart for Availability
€ 82,70

About This Item

Linear Formula:
CH3COOCH2CH=CH2
CAS Number:
Molecular Weight:
100.12
UNSPSC Code:
12352108
EC Index Number:
209-734-8
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
Form:
liquid

€ 82,70


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

27.2 hPa ( 20 °C)

Quality Segment

assay

≥98.0% (GC)

form

liquid

autoignition temp.

374 °C

potency

130 mg/kg LD50, oral (Rat), 1021 mg/kg LD50, skin (Rabbit)

expl. lim.

2.1-13.0 % (v/v)

bp

103-104 °C/1013 mbar

mp

-96 °C

transition temp

flash point 6 °C

storage temp.

2-30°C

SMILES string

O(CC=C)C(=O)C

InChI

1S/C5H8O2/c1-3-4-7-5(2)6/h3H,1,4H2,2H3

InChI key

FWZUNOYOVVKUNF-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.185598.14990PHR3189
assay

≥98.0% (GC)

assay

≥99.0% (GC)

assay

≥98.0% (GC)

assay

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

300

form

liquid

form

liquid

form

liquid

form

-

mp

-96 °C

mp

-40.0 °C

mp

-

mp

−99 °C (lit.)

bp

103-104 °C/1013 mbar

bp

89-90 °C/5 hPa

bp

-

bp

115-117 °C (lit.)

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C


Still not finding the right product?

Explore all of our products under Allyl acetate


pictograms

FlameSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters