Skip to Content
Merck

Skip To

8.20366

trans-1,2-Cyclohexanediol

for synthesis

Synonym(s):

trans-1,2-Cyclohexanediol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 g
Check Cart for Availability
€ 94,30
€ 70,72

About This Item

Linear Formula:
C6H10-1,2-(OH)2
CAS Number:
Molecular Weight:
116.16
UNSPSC Code:
12352104
EC Index Number:
215-956-6
NACRES:
NA.22
MDL number:

€ 70,72

List Price€ 94,30Save 25%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

4 hPa ( 110 °C)

Quality Segment

assay

≥98.0% (GC)

form

powder

autoignition temp.

360 °C

bp

231-233 °C

mp

100-103 °C (External MSDS)

transition temp

flash point 134 °C (External MSDS)

storage temp.

2-30°C

SMILES string

O[C@H]1[C@@H](CCCC1)O

InChI

1S/C6H12O2/c7-5-3-1-2-4-6(5)8/h5-8H,1-4H2/t5-,6-/m1/s1

InChI key

PFURGBBHAOXLIO-PHDIDXHHSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 100 °C
Melting range (upper value): ≤ 104 °C
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1417128.41088560286
assay

≥98.0% (GC)

assay

98%

assay

≥97.0% (acidimetric)

assay

-

storage temp.

2-30°C

storage temp.

-

storage temp.

2-30°C

storage temp.

-

bp

231-233 °C

bp

-

bp

-

bp

-

form

powder

form

solid

form

powder

form

-

mp

100-103 °C (External MSDS)

mp

101-104 °C (lit.)

mp

220-221 °C

mp

160 °C (lit.)

autoignition temp.

360 °C

autoignition temp.

-

autoignition temp.

-

autoignition temp.

-


Still not finding the right product?

Explore all of our products under trans-1,2-Cyclohexanediol


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
820366002504022536457420

Questions

Reviews

No rating value

Active Filters