Skip to Content
Merck

Skip To

8.20243

Di-tert-butyl disulfide

for synthesis

Synonym(s):

Di-tert-butyl disulfide, tert-Butyl disulfide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 mL
Check Cart for Availability
€ 207,00

About This Item

Empirical Formula (Hill Notation):
C8H18S2
CAS Number:
Molecular Weight:
178.36
UNSPSC Code:
12352100
EC Index Number:
203-734-1
NACRES:
NA.22
MDL number:
Assay:
≥95.0% (GC)
Form:
liquid

€ 207,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

2 hPa ( 38 °C)

Quality Segment

assay

≥95.0% (GC)

form

liquid

potency

6200 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rabbit)

expl. lim.

0.8 % (v/v)

refractive index

n/D 1.489-1.491 (N20/D)

mp

-5 °C

transition temp

flash point 67 °C

density

0.93 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

S(SC(C)(C)C)C(C)(C)C

InChI

1S/C8H18S2/c1-7(2,3)9-10-8(4,5)6/h1-6H3

InChI key

BKCNDTDWDGQHSD-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)

Refractive index (N20/D) : 1.489 – 1.491

Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.20242419362124311
assay

≥95.0% (GC)

assay

≥97.0% (GC)

assay

95%

assay

98%

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

mp

-5 °C

mp

-94 °C

mp

-

mp

-

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

-

form

liquid

form

liquid

form

liquid

form

liquid

refractive index

n/D 1.489-1.491 (N20/D)

refractive index

-

refractive index

n20/D 1.444 (lit.)

refractive index

n20/D 1.45 (lit.)


Still not finding the right product?

Explore all of our products under Di-tert-butyl disulfide


pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

143.6 °F - closed cup

flash_point_c

62 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters