Skip to Content
Merck

Skip To

8.14902

2-Amino-2-cyanoacetamide

for synthesis

Synonym(s):

2-Amino-2-cyanoacetamide, ACA

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C3H5N3O
CAS Number:
Molecular Weight:
99.09
UNSPSC Code:
12352111
EC Index Number:
229-771-3
Form:
powder


Quality Segment

form

powder

potency

>2000 mg/kg LD50, oral (Rat)

mp

119-122 °C

InChI

1S/C3H5N3O/c4-1-2(5)3(6)7/h2H,5H2,(H2,6,7)

InChI key

JRWAUKYINYWSTA-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 97.0 %
Melting range (lower value): ≥ 120 °C
Melting range (upper value): ≤ 124 °C
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under 2-Amino-2-cyanoacetamide


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters