Skip to Content
Merck
All Photos(2)

Key Documents

8.08742

Sigma-Aldrich

N,N,N′,N′-Tetramethyl ethylenediamine

for synthesis

Synonym(s):

N,N,N′,N′-Tetramethyl ethylenediamine, TEMED, TMEDA, 1,2-Bis(dimethylamino)ethane

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€28.40
250 ML
€48.80
1 L
€124.00

€28.40


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€28.40
250 ML
€48.80
1 L
€124.00

About This Item

Linear Formula:
(CH3)2NCH2CH2N(CH3)2
CAS Number:
Molecular Weight:
116.20
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-744-6
NACRES:
NA.22

€28.40


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

21 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

145 °C

potency

5390 mg/kg LD50, skin (Rabbit)

expl. lim.

1-9 % (v/v)

pH

8.0-8.5 (20 °C, 0.1 g/L in H2O)

bp

121 °C/1013 hPa

mp

-55 °C

transition temp

flash point 19 °C

density

0.78 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCN(C)C)(C)C

InChI

1S/C6H16N2/c1-7(2)5-6-8(3)4/h5-6H2,1-4H3

InChI key

KWYHDKDOAIKMQN-UHFFFAOYSA-N

Application


  • Surface Basicity and Hydrophilic Character of Coal Ash-Derived Zeolite NaP1 Modified by Fatty Acids: This study discusses the modification of Zeolite NaP1 with N,N,N′,N′-tetramethylethylenediamine to assess changes in surface basicity and hydrophilic character, relevant for environmental chemistry and materials science (AP Beltrão-Nunes, M Pires, R Roy, A Azzouz, 2024).

  • Horseradish peroxidase-catalyzed polyacrylamide gels: Explores the use of N,N,N′,N′-tetramethylethylenediamine in the polymerization of polyacrylamide gels, enhancing the understanding of gel-based biomolecular analysis systems (C Liao, T Li, F Chen, S Yan, L Zhu, H Tang, D Wang, 2024).

  • Photoinduced Alkylsulfonylation and Cyanoalkylsulfonylation of Morita–Baylis–Hillman Adducts: Demonstrates the role of N,N,N′,N′-tetramethylethylenediamine in photochemical synthesis, providing insights into organic synthesis mechanisms (ZJ Song, Y Bao, YJ Sun, S Yan, Y Zhang, 2024).

  • Probing coordination geometry and electronic nature of Cu center in mixed ligand copper complexes: This research utilizes N,N,N′,N′-tetramethylethylenediamine to study the coordination geometry and electronic properties of copper complexes, significant for inorganic chemistry and catalysis (M Bairagi, VK Hinge, N Yadav, SK Joshi, 2024).

  • Indirect Adhesion of Hydrogels via the Radical Polymerization: Investigates the use of N,N,N′,N′-tetramethylethylenediamine in creating an indirect adhesive system for hydrogels, relevant to material science and polymer chemistry (R Okada, S Tamesue, 2024).


Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.774 - 0.777
Water (K. F.): ≤ 0.50 %
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

61.7 °F - closed cup

Flash Point(C)

16.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service