Skip to Content
Merck
All Photos(1)

Key Documents

1.01200

Supelco

Ammonium peroxodisulfate

EMPLURA®

Synonym(s):

Ammonium peroxodisulfate, Ammonium persulfate, Peroxidisulfuric acid diammonium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
€70.20
5 KG
€246.00

€70.20


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
1 KG
€70.20
5 KG
€246.00

About This Item

Linear Formula:
(NH4)2S2O8
CAS Number:
Molecular Weight:
228.20
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-786-5
NACRES:
NA.21

€70.20


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

product line

EMPLURA®

Assay

≥98.0% (iodometric)

form

solid

potency

742 mg/kg LD50, oral (Rat)

ign. residue

≤0.1% (as SO4)

pH

3.2 (20 °C, 100 g/L in H2O)

mp

120 °C (decomposition)

solubility

620 g/L

density

1.98 g/cm3 at 20 °C

bulk density

900 kg/m3

anion traces

chloride (Cl-): ≤0.002%

cation traces

Fe: ≤0.001%
Mn: ≤0.0002%
heavy metals (as Pb): ≤0.003%

storage temp.

15-25°C

SMILES string

[S](=O)(=O)([O-])OO[S](=O)(=O)[O-].[N+H4].[N+H4]

InChI

1S/2H3N.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h2*1H3;(H,1,2,3)(H,4,5,6)

InChI key

ROOXNKNUYICQNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Novel Hydrolytic Degradable Crosslinked Interpenetrating Polymeric Networks (IPNs): An Efficient Hybrid System to Manage the Controlled Release and Degradation of Misoprostol.: This study explores the use of ammonium peroxodisulfate as an initiator in the synthesis of hydrolytically degradable IPNs for controlled drug release. The innovative approach provides a new avenue for drug delivery systems in pharmaceutical applications (Mehmood et al., 2023).

  • Amikacin-Loaded Chitosan Hydrogel Film Cross-Linked with Folic Acid for Wound Healing Application.: The research utilizes ammonium peroxodisulfate to cross-link chitosan hydrogel films loaded with amikacin, demonstrating enhanced antibacterial properties and potential for wound healing applications (Mehmood et al., 2023).

  • Synthesis and In Vitro Evaluation of Aspartic Acid Based Microgels for Sustained Drug Delivery.: This paper highlights the role of ammonium peroxodisulfate in the synthesis of aspartic acid-based microgels, which exhibit promising results for sustained drug delivery systems (Suhail et al., 2021).

  • Fabrication and In Vitro Evaluation of pH-Sensitive Polymeric Hydrogels as Controlled Release Carriers.: The study investigates the use of ammonium peroxodisulfate in the fabrication of pH-sensitive polymeric hydrogels, offering potential for controlled release applications in pharmaceuticals (Suhail et al., 2021).

  • Extraction of Lithium from Single-Crystalline Lithium Manganese Oxide Nanotubes Using Ammonium Peroxodisulfate.: This research presents a novel method for extracting lithium using ammonium peroxodisulfate, highlighting its significance in the development of advanced materials for energy storage applications (Shi et al., 2020).


Analysis Note

Assay (iodometric): ≥ 98.0 %
Identity: passes test
Chloride (Cl): ≤ 0.002 %
Heavy metals (as Pb): ≤ 0.003 %
Fe (Iron): ≤ 0.001 %
Mn (Manganese): ≤ 0.0002 %
Residue on ignition (as SO₄): ≤ 0.1 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service