1.10732
N,N,N′,N′-Tetramethyl ethylenediamine (Temed)
GR for analysis
Synonym(s):
N,N,N′,N′-Tetramethyl ethylenediamine (Temed), TEMED, TMEDA, 1,2-Bis(dimethylamino)ethane
About This Item
Recommended Products
vapor pressure
21 hPa ( 20 °C)
Quality Level
Assay
≥99% (GC)
form
liquid
autoignition temp.
145 °C
potency
5390 mg/kg LD50, skin (Rabbit)
expl. lim.
1-9 % (v/v)
pH
8.0-8.5 (20 °C, 0.1 g/L in H2O)
bp
121 °C/1013 hPa
mp
-55 °C
transition temp
flash point 19 °C
density
0.78 g/cm3 at 20 °C
storage temp.
15-25°C
InChI
1S/C6H16N2/c1-7(2)5-6-8(3)4/h5-6H2,1-4H3
InChI key
KWYHDKDOAIKMQN-UHFFFAOYSA-N
Related Categories
Analysis Note
Identity (IR-spectrum): passes test
Fe (Iron): ≤ 0.00001 %
Sulfated ash: ≤ 0.005 %
Water: ≤ 0.5 %
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
61.7 °F - closed cup
Flash Point(C)
16.5 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service