Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 50 mL | Check Cart for Availability | € 45,00 |
About This Item
Linear Formula:
CH3(CH2)7CHO
CAS Number:
Molecular Weight:
142.24
UNSPSC Code:
12352114
EC Index Number:
204-688-5
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
€ 45,00
Check Cart for Availability
Quality Segment
assay
≥98.0% (GC)
potency
>5000 mg/kg LD50, oral (Rat)
transition temp
flash point 76 °C
density
0.83 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
O=CCCCCCCCC
InChI
1S/C9H18O/c1-2-3-4-5-6-7-8-9-10/h9H,2-8H2,1H3
InChI key
GYHFUZHODSMOHU-UHFFFAOYSA-N
Analysis Note
Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.823 - 0.827
Identity (IR): passes test
Density (d 20 °C/ 4 °C): 0.823 - 0.827
Identity (IR): passes test
Still not finding the right product?
Explore all of our products under Nonanal
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 3 - Skin Irrit. 2
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
147.2 °F - closed cup
flash_point_c
64 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 8071660050 | 04022536394282 |
