Skip to Content
Merck
All Photos(1)

Key Documents

8.05930

Sigma-Aldrich

Manganese(II) chloride

for synthesis

Synonym(s):

Manganese(II) chloride, Manganese dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€38.40
500 G
€120.00

€38.40


Estimated to ship on28 March 2025


Request a Bulk Order

Select a Size

Change View
100 G
€38.40
500 G
€120.00

About This Item

Empirical Formula (Hill Notation):
Cl2Mn
CAS Number:
Molecular Weight:
125.84
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-869-6
NACRES:
NA.22
form:
scales

€38.40


Estimated to ship on28 March 2025


Request a Bulk Order

Quality Level

form

scales

potency

250 mg/kg LD50, oral (Rat)

pH

5.5 (20 °C, 25 g/L in H2O)

bp

1190 °C/1013 hPa

mp

650 °C

density

2.977 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

[Mn+2].[Cl-].[Cl-].O.O.O.O

InChI

1S/2ClH.Mn.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

CNFDGXZLMLFIJV-UHFFFAOYSA-L

Analysis Note

Assay (out of Mn): ≥ 96 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Brain

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service