Skip to Content
Merck

Skip To

1.12237

Manganese

powder 99+

Synonym(s):

Manganese

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 g
Check Cart for Availability
€ 94,00

About This Item

Empirical Formula (Hill Notation):
Mn
CAS Number:
Molecular Weight:
54.94
UNSPSC Code:
12141724
EC Index Number:
231-105-1
NACRES:
NA.22
MDL number:

€ 94,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

potency

9000 mg/kg LD50, oral (Rat)

bp

2061 °C

mp

1246 °C

density

7.4 g/cm3 at 19 °C

storage temp.

no temp limit

SMILES string

[Mn]

InChI

1S/Mn

InChI key

PWHULOQIROXLJO-UHFFFAOYSA-N

Analysis Note

Identity (elemental analysis): passes test
Assay (complexometric): ≥ 99.0 %
Particle size (< 100 µm): ≥ 90 %

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
463728GF47272686GF01168736
Quality Level

200

Quality Level

100

Quality Level

-

Quality Level

-

storage temp.

no temp limit

storage temp.

-

storage temp.

-

storage temp.

-

mp

1246 °C

mp

1244 °C (lit.)

mp

1244 °C (lit.)

mp

1244 °C (lit.)

density

7.4 g/cm3 at 19 °C

density

7.3 g/mL at 25 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

bp

2061 °C

bp

1962 °C (lit.)

bp

1962 °C (lit.)

bp

1962 °C (lit.)

potency

9000 mg/kg LD50, oral (Rat)

potency

-

potency

-

potency

-


Still not finding the right product?

Explore all of our products under Manganese


pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Sol. 2

Storage Class

4.1B - Flammable solid hazardous materials

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters