Skip to Content
Merck
All Photos(1)

Key Documents

429449

Sigma-Aldrich

Manganese(II) chloride

AnhydroBeads, −10 mesh, 99.99% trace metals basis

Synonym(s):

Manganese dichloride, Sacchite

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€75.20
25 G
€285.00

€75.20


Estimated to ship on29 March 2025


Request a Bulk Order

Select a Size

Change View
5 G
€75.20
25 G
€285.00

About This Item

Linear Formula:
MnCl2
CAS Number:
Molecular Weight:
125.84
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
form:
beads

€75.20


Estimated to ship on29 March 2025


Request a Bulk Order

product line

AnhydroBeads

Quality Level

Assay

99.99% trace metals basis

form

beads

impurities

≤150.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

652 °C (lit.)

density

2.98 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Cl[Mn]Cl

InChI

1S/2ClH.Mn/h2*1H;/q;;+2/p-2

InChI key

GLFNIEUTAYBVOC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Manganese(II) chloride is an inorganic salt soluble in water. It exists in several hydrated forms, including the anhydrous, dihydrate (MnCl2.2H2O), and tetrahydrate (MnCl24H2O) states. It is used in the production of dry cell batteries and as a precursor for other manganese compounds. Additionally, manganese(II) chloride is also employed in the synthesis of organomanganese compounds and used as a catalyst in various organic reactions.

Application

Manganese(II) chloride can be used as:      
  • A precursor in the preparation of manganese oxide coated natural spherical graphite materials, which are used for improving the electrochemical performance of the graphite materials, making them suitable for applications in batteries and supercapacitors.     
  • A key component in the preparation of polyvinyl alcohol/manganese chloride composite films, which are find potential applications in areas such as sensors, energy storage devices, and flexible electronics due to their improved optical, mechanical, and thermal properties.      
  • A precursor in the synthesis of Mn-doped CsPbCl3 perovskite nanocrystals for enhancing their optical properties and quantum yield, which are vital for their application in improving the efficiency of solar cells.  
  • A catalyst for the transfer dehydrogenation of glycerol to lactic acid.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Brain

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isaac A Adedara et al.
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS), 39, 21-29 (2016-12-03)
Despite the fact that most environmental exposures to metals do not occur in isolation, the combined effects of metal mixtures on brain-pituitary-gonadal axis are poorly known. The present study investigated the impacts of co-exposure to arsenic (As) and manganese (Mn)
Bingxing Zhang et al.
Nature communications, 10(1), 2980-2980 (2019-07-07)
Developing highly efficient electrocatalysts based on cheap and earth-abundant metals for CO2 reduction is of great importance. Here we demonstrate that the electrocatalytic activity of manganese-based heterogeneous catalyst can be significantly improved through halogen and nitrogen dual-coordination to modulate the
Abbati, G.L. et al.
Inorganic Chemistry, 37, 1430-1430 (1998)
Tang, J. et al.
Tetrahedron, 55, 1893-1893 (1999)
Marta Sidoryk-Wegrzynowicz et al.
Journal of neurochemistry, 122(4), 856-867 (2012-06-20)
Manganese (Mn) has been implicated in the impairment of the glutamate-glutamine cycling (GGC) by deregulation of Glu and glutamine (Gln) turnover in astrocytes. Here, we have examined possible mechanisms involved in the Mn(II)-mediated disruption of Glu turnover, including those related

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Questions

  1. How can a glass ampule-like vial be opened without a score mark?

    1 answer
    1. Using a diamond glass cutter like item 933392 or a sharp-edged triangle metal file is recommended to score the vial all around in order to break it open.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service