Skip to Content
Merck

Skip To

8.04166

2-Bromophenol

for synthesis

Synonym(s):

2-Bromophenol, 2-Bromo-1-hydroxybenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 mL
Check Cart for Availability
€ 138,00

About This Item

Empirical Formula (Hill Notation):
C6H5BrO
CAS Number:
Molecular Weight:
173.01
UNSPSC Code:
12352104
EC Index Number:
202-432-7
NACRES:
NA.22
MDL number:
Form:
liquid

€ 138,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

liquid

bp

195 °C

mp

5 °C

transition temp

flash point 108 °C

density

1.63 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Brc1c(cccc1)O

InChI

1S/C6H5BrO/c7-5-3-1-2-4-6(5)8/h1-4,8H

InChI key

VADKRMSMGWJZCF-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.02253750778B75808
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

density

1.63 g/cm3 at 20 °C

density

1.26 g/cm3 at 20 °C

density

-

density

-

bp

195 °C

bp

174 °C/1013 hPa

bp

-

bp

235-236 °C (lit.)

mp

5 °C

mp

7 °C

mp

96-103 °C

mp

61-64 °C (lit.)

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

-

form

liquid

form

liquid

form

solid

form

solid


Still not finding the right product?

Explore all of our products under 2-Bromophenol


signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

107.6 °F - closed cup

flash_point_c

42 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
804166999904027269034306
804166002504027269035587

Questions

Reviews

No rating value

Active Filters