Skip to Content
Merck

Skip To

8.01556

Isobutyraldehyde

for synthesis

Synonym(s):

Isobutyraldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Check Cart for Availability
€ 31,10
1 L
Check Cart for Availability
€ 45,20
20 kg
Check Cart for Availability
€ 494,00

About This Item

Linear Formula:
(CH3)2CHCHO
CAS Number:
Molecular Weight:
72.11
UNSPSC Code:
12352114
EC Index Number:
201-149-6
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
Form:
liquid

€ 31,10


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

189 hPa ( 20 °C)

Quality Segment

assay

≥98.0% (GC)

form

liquid

autoignition temp.

165 °C

potency

>3000 mg/kg LD50, oral (Rat), >5000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.6-11.0 % (v/v)

bp

63-64.5 °C/1013 mbar

mp

-65.9 °C

transition temp

flash point -20 °C

density

0.79 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=CC(C)C

InChI

1S/C4H8O/c1-4(2)3-5/h3-4H,1-2H3

InChI key

AMIMRNSIRUDHCM-UHFFFAOYSA-N

Features and Benefits

Saturation Concentration in Air: 452 g/m3

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.788 - 0.790
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
685291347880W222011
assay

≥98.0% (GC)

assay

≥97.0% (GC)

assay

-

assay

96%

Quality Level

200

Quality Level

100

Quality Level

-

Quality Level

300, 400

bp

63-64.5 °C/1013 mbar

bp

63 °C (lit.)

bp

63 °C (lit.)

bp

63 °C (lit.)

form

liquid

form

-

form

-

form

-

mp

-65.9 °C

mp

−65 °C (lit.)

mp

−65 °C (lit.)

mp

−65 °C (lit.)

density

0.79 g/cm3 at 20 °C

density

0.79 g/mL at 25 °C (lit.)

density

0.79 g/mL at 25 °C (lit.)

density

0.79 g/mL at 25 °C (lit.)


Still not finding the right product?

Explore all of our products under Isobutyraldehyde


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

-11.2 °F - closed cup

flash_point_c

-24 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters