Skip to Content
Merck
All Photos(1)

Key Documents

8.00472

Sigma-Aldrich

Isobutyric acid

for synthesis

Synonym(s):

Isobutyric acid, 2-Methylpropionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€37.30
1 L
€63.80

€37.30


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€37.30
1 L
€63.80

About This Item

Linear Formula:
(CH3)2CHCOOH
CAS Number:
Molecular Weight:
88.11
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-195-7
NACRES:
NA.22

€37.30


Estimated to ship on22 May 2025


Request a Bulk Order

vapor pressure

2 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

455 °C

potency

266 mg/kg LD50, oral (Rat)
475 mg/kg LD50, skin (Rabbit)

expl. lim.

1.6-7.3 % (v/v)

pH

2.3 (25 °C, 500 g/L in H2O)

mp

-46 °C

transition temp

flash point 62 °C

solubility

618 g/L

density

0.95 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OC(=O)C(C)C

InChI

1S/C4H8O2/c1-3(2)4(5)6/h3H,1-2H3,(H,5,6)

InChI key

KQNPFQTWMSNSAP-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.947 - 0.949
Water (K. F.): ≤ 0.20 %
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service