Skip to Content
Merck
All Photos(1)

Key Documents

58360

Sigma-Aldrich

Isobutyric acid

puriss. p.a., ≥99.5%

Synonym(s):

2-Methylpropionic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€35.50

€35.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€35.50

About This Item

Linear Formula:
(CH3)2CHCO2H
CAS Number:
Molecular Weight:
88.11
Beilstein:
635770
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€35.50


Check Cart for Availability

Request a Bulk Order

vapor density

3.04 (vs air)

Quality Level

vapor pressure

1.5 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥99.5% (GC)
≥99.5%

autoignition temp.

824 °F

expl. lim.

10 %

impurities

≤0.2% water

refractive index

n20/D 1.393 (lit.)
n20/D 1.393

bp

153-154 °C (lit.)

mp

−47 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤2 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

functional group

carboxylic acid

SMILES string

CC(C)C(O)=O

InChI

1S/C4H8O2/c1-3(2)4(5)6/h3H,1-2H3,(H,5,6)

InChI key

KQNPFQTWMSNSAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isobutyric acid can be prepared via the carbonylation of propylene.[1]

Application

Isobutyric acid may be used to synthesize methacrylic acid via oxidative dehydrogenation in the presence of a suitable heteropolyacid as catalyst.[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Acidic cesium salts of molybdovanadophosphoric acids as efficient catalysts for oxidative dehydrogenation of isobutyric acid"
Lee YK, et al.
Catalysis Today, 33(1-3), 183-189 (1997)
Kimberlee S Mix et al.
Molecular pharmacology, 65(2), 309-318 (2004-01-27)
Matrix metalloproteinases (MMPs) degrade extracellular matrix components, and overexpression of these enzymes contributes to tissue destruction in arthritis. Of particular importance are the collagenases, MMP-1 and MMP-13, which have high activity against the interstitial collagens in cartilage. In this study
Nicoletta Beffagna et al.
Journal of experimental botany, 58(15-16), 4183-4194 (2007-11-28)
In Arabidopsis thaliana cells, fusicoccin (FC) treatment induced an early and marked increase in the extracellular H(2)O(2) level. It also increased the huge hypo-osmotic stress-induced oxidative wave and, in addition, prevented the H(2)O(2) peak drop. These effects were apparently not
Julia Biermann et al.
Molecular vision, 17, 395-403 (2011-02-12)
Histone deacetylase inhibitors (HDACi) have neuroprotective effects under various neurodegenerative conditions, e.g., after optic nerve crush (ONC). HDACi-mediated protection of central neurons by increased histone acetylation has not previously been demonstrated in rat retinal ganglion cells (RGCs), although epigenetic changes
A Vyalikh et al.
Physical chemistry chemical physics : PCCP, 9(18), 2249-2257 (2007-05-10)
Solid state deuterium NMR has been used to study the molecular motion of d(6)-isobutyric acid (d(6)-iBA) in the pure (unconfined) state and confined in the cylindrical pores of two periodic mesoporous silica materials (MCM-41, pore size 3.3 nm and SBA-15

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service