Skip to Content
Merck

Skip To

79130BC

GDS PPMX Handheld Barcode Reader

BioControl, For use with GDS PickPen® PIPETMAX®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 ea
Check Cart for Availability
€ 1.520,00

About This Item

UNSPSC Code:
41121521
NACRES:
NC.81

€ 1.520,00


Check Cart for Availability

Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

For use with GDS PICKPEN™ PIPETMAX™ System

manufacturer/tradename

BioControl Systems 79130

storage condition

room temperature

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with BioControl GDS

storage temp.

room temp

Application

PCR, Immunomagnetic Separation (IMS)

Other Notes

One 2D barcode reader

Legal Information

PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
79129BC79133BC79155BC
compatibility

for use with BioControl GDS

compatibility

BioControl GDS

compatibility

for use with GDS PickPen® PIPETMAX System

compatibility

for use with GDS PickPen® PIPETMAX System

storage condition

room temperature

storage condition

room temperature

storage condition

-

storage condition

-

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

manufacturer/tradename

BioControl Systems 79130

manufacturer/tradename

BioControl Systems 79129

manufacturer/tradename

BioControl Systems

manufacturer/tradename

BioControl Systems




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters