Skip to Content
Merck

Skip To

79115BC

GDS PickPen® PIPETMAX® Power Supply

BioControl Systems, for use with GDS PickPen® PIPETMAX, EU adaptor

Synonym(s):

GDS PPMX Power Supply, GDS Pipetmax® Power Source

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 ea
Check Cart for Availability
€ 513,00

About This Item

UNSPSC Code:
41121521
NACRES:
NC.81

€ 513,00


Check Cart for Availability

Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pkg of 1 unit

manufacturer/tradename

BioControl Systems

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS PickPen® PIPETMAX, EU adaptor

storage temp.

room temp

General description

GDS PickPen® PIPETMAX® (PPMX) Power Supply is a power supply unit designed specifically for use with the GDS PPMX automated sample preparation system within the EU. It provides electrical power to the PPMX instrument, enabling it to perform automated liquid handling and IMS operations. The GDS PPMX Power Supply ensures a reliable and consistent power supply to support the functionality of the GDS system.

Application

GDS PPMX Power Supply is used with GDS PickPen® PIPETMAX® to complete sample preparation for the detection of pathogenic organisms in foods, ingredients, and environmental samples.

Legal Information

PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
79113BC79112BC79114BC
compatibility

for use with GDS PickPen® PIPETMAX, EU adaptor

compatibility

for use with GDS PickPen® PIPETMAX, EU adaptor

compatibility

for use with GDS PickPen® PIPETMAX System

compatibility

for use with GDS PickPen® PIPETMAX, UK adaptor

packaging

pkg of 1 unit

packaging

-

packaging

-

packaging

-

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

manufacturer/tradename

BioControl Systems

manufacturer/tradename

BioControl Systems

manufacturer/tradename

BioControl Systems

manufacturer/tradename

BioControl Systems 79114

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters