Skip to Content
Merck
All Photos(1)

Key Documents

1.09435

Supelco

Buffer solution

(citric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 4.00 (20°C) Certipur®

Synonym(s):

Buffer solution pH 4.00 (20°C) (citric acid/sodium hydroxide/hydrogen chloride)

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€67.40
4 L
€226.00
10 L
€325.00

€67.40


Estimated to ship on25 March 2025


Request a Bulk Order

Select a Size

Change View
1 L
€67.40
4 L
€226.00
10 L
€325.00

About This Item

UNSPSC Code:
12161704
NACRES:
NA.25

€67.40


Estimated to ship on25 March 2025


Request a Bulk Order

grade

certified reference material

Quality Level

product line

Certipur®

pH

4.0 (20 °C in H2O)

density

1.01 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The buffer solution (citric acid/sodium hydroxide/hydrogen chloride) is gravimetrically prepared using high-quality reagents in high-purity water. It is quality controlled and qualified in an ISO/IEC 17025 accredited calibration lab. Its pH value of 4.00 at 20°C is qualified using a combined glass electrode following a 5-point calibration method, in accordance with DIN 19268, against certified primary reference standards as per DIN 19266, IUPAC, NIST, USP, and Ph. Eur.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

It is used in analytical procedures for calibration, qualification, and monitoring of pH measuring instruments and as a control for testing other buffer solutions.

Features and Benefits

  • Measured and qualified in an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to SRMs from NIST and primary certified reference standards from PTB
  • Ready-to-use solution available in a sealed bottle and an innovative Titripac® packaging for positive environmental impact due to reduced waste generation
  • Long-lasting with a shelf life of 3 years
  • Accompanied with a batch-specific and detailed certificate of analysis (CoA) for an effective audit

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Titripac is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

recommended

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Regarding the pH buffer standards, after opening, can I reuse the solution more than once to check the pH?

    1 answer
    1. Once the buffer aliquot has been poured out of the bottle, it can only be used once. And make sure not to pour it back into the bottle.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service