Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 20 μg | Check Cart for Availability | € 487,00 |
About This Item
NACRES:
NA.51
UNSPSC Code:
41106611
€ 487,00
Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1830
form
liquid
Quality Segment
manufacturer/tradename
Novagen®
storage condition
OK to freeze
shipped in
wet ice
storage temp.
−20°C
General description
The pBiEx vectors are designed for cloning and high-level expression of proteins in both bacteria and in transiently transfected Spodoptera-derived insect cells. In E. coli, target gene transcription is regulated by the tightly controlled T7lac promoter, and a ribosome binding site (rbs) directs translation initiation. In insect cells, transcription is driven by the AcNPV-derived hr5 enhancer and immediate early promoter, IE1.
pBiEx-1 contains N-terminal His•Tag and S•Tag coding sequences, and a C-terminal HSV•Tag coding sequence. For sequencing or PCR of recombinants in pBiEx-1, refer to the table below to choose the appropriate primers.
Commercial use of this Product requires a commercial license from EMD Millipore Corporation. Commercial use shall include but not be limited to (1) use of the Product or its components in manufacturing; (2) use of the Product or its components to provide a service, information, or data to others in exchange for consideration; (3) use of the Product or its components (or any derivatives thereof) for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the Product or its components, whether or not such Product or its components are resold for use in research. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of this Product.
pBiEx-1 contains N-terminal His•Tag and S•Tag coding sequences, and a C-terminal HSV•Tag coding sequence. For sequencing or PCR of recombinants in pBiEx-1, refer to the table below to choose the appropriate primers.
Commercial use of this Product requires a commercial license from EMD Millipore Corporation. Commercial use shall include but not be limited to (1) use of the Product or its components in manufacturing; (2) use of the Product or its components to provide a service, information, or data to others in exchange for consideration; (3) use of the Product or its components (or any derivatives thereof) for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the Product or its components, whether or not such Product or its components are resold for use in research. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of this Product.
Legal Information
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
This product is sold for internal research use only. Any commercial use of this product, its components, and/or any derivatives thereof (including but not limited to proteins produced using the product or its components) (together and hereinafter the “EMD Product”) requires signature of a written commercial use agreement with EMD Millipore Corporation or its successor-in-interest. Commercial use shall include but not be limited to: (1) use of the EMD Product to manufacture products for sale to third parties; (2) use of the EMD Product to provide services, information, or data to third parties in exchange for consideration; (3) use of the EMD Product for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the EMD Product, whether or not such EMD Product is resold for research use. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of the EMD Product. Please direct any questions on these use restrictions to: [email protected].
Toxicity: Standard Handling (A)
1 of 1
This Item | |||
|---|---|---|---|
| form liquid | form liquid | form liquid | form liquid |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| storage temp. −20°C | storage temp. −70°C | storage temp. −20°C | storage temp. −20°C |
| storage condition OK to freeze | storage condition OK to freeze | storage condition OK to freeze | storage condition OK to freeze |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| manufacturer/tradename Novagen® | manufacturer/tradename Novagen® | manufacturer/tradename Novagen® | manufacturer/tradename Novagen® |
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 71234-3 | 04055977271232 |


