Skip to Content
Merck

Skip To

70955

Casein

5%, Alkali-soluble, Unique blocking reagent for ultra-low background

Synonym(s):

blocking buffer, immunodetection blocking buffer

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41105329
NACRES:
NB.23
Form:
liquid


Product Name

5% Alkali-soluble Casein, Unique blocking reagent for ultra-low background

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

Alkali-Soluble Casein is a superior blocking reagent for Western blot applications. This preparation minimizes non-specific background relative to other blocking reagents, such as BSA, gelatin, or non-fat milk. Alkali-Soluble Casein is effective in colorimetric and chemiluminescent detection protocols. Note that due to its high coating properties, detection sensitivity for some antibodies may be lower than with other blocking reagents under the same conditions (although the background is also reduced); the concentration can be optimized for maximal signal:noise ratios. Supplied as a 5% w/v solution; recommended working concentration range is 0.251%.
Unique blocking reagent for ultra-low background

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C7594B6429WBAVDCH01
form

liquid

form

powder blend

form

solution

form

-

storage condition

OK to freeze

storage condition

-

storage condition

-

storage condition

-

shipped in

ambient

shipped in

-

shipped in

-

shipped in

ambient

storage temp.

2-8°C

storage temp.

room temp

storage temp.

room temp

storage temp.

15-25°C

manufacturer/tradename

Novagen®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

Immobilon®


Still not finding the right product?

Explore all of our products under 5% Alkali-soluble Casein


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Protocols

Protokoll für die Probenvorbereitung zur Zelllyse und effizienten Proteinextraktion aus kultivierten Geweben und Zellen für das anschließende Western Blotting.

Protocol for sample preparation for cell lysis and efficient protein extraction from cultured tissues and cells for subsequent Western blotting.





Global Trade Item Number

SKUGTIN
70955-304055977256161

Questions

  1. Can I have a detailed composition of the product?

    1 answer
    1. Unfortunately, the exact formulation is proprietary, however, it is simply a 5% Casein solution in 5X TBS. It does include a preservative at low concentrations.

      Helpful?

Reviews

No rating value

Active Filters