Skip to Content
Merck
All Photos(1)

Documents

70796-M

Sigma-Aldrich

His•Tag® Monoclonal Antibody

Novagen®

Synonym(s):

Anti-His antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

antibody form

purified antibody

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

storage temp.

2-8°C

target post-translational modification

unmodified

Related Categories

General description

Sensitive, specific detection of His•Tag fusion proteins
The HIS TAG® Monoclonal Antibody is a mouse monoclonal antibody (IgG1) directed against the HIS TAG® sequence encoded by many of the NOVAGEN® expression vectors as well as many other commercially available vectors. The antibody recognizes five consecutive histidine residues regardless of the surrounding amino acid context. The high affinity (Kd = 5 × 10-8 to 1 × 10-9 M) enables sensitive, specific detection of HIS TAG® fusion proteins at antibody concentrations of 0.1-0.2 g/ml. The HIS TAG® Monoclonal Antibody binds to N-terminal, C-terminal, and internal HIS TAG® sequences. This antibody also detects the recombinant marker bands in the Perfect Protein<TMSYMBOL></TMSYMBOL> and Trail Mix<TMSYMBOL></TMSYMBOL> Western Markers for convenient visualization of accurate internal standards on Western blots*. For lowest background in Western blotting applications, Alkali-soluble Casein is recommended as a blocking agent. The 100-g package size provides enough purified antibody for 1000 mL of working solution.

* Not recommended for HRP-based detection of Trail Mix Western Markers.

Warning

Toxicity: Standard Handling (A)

Legal Information

HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chia-Yang Chen et al.
The Plant journal : for cell and molecular biology, 112(6), 1462-1472 (2022-11-12)
Histone deacetylases (HDAs) regulate many aspects of plant development and responses to environmental changes. Previous studies have demonstrated that the Arabidopsis histone deacetylase HDA15 is a positive regulator in far-red (FR) light-mediated inhibition of hypocotyl elongation. Furthermore, HDA15 can be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service