Skip to Content
Merck

Skip To

70541

GST•Bind Resin

Synonym(s):

GST Resin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
10 mL
Check Cart for Availability
€ 314,00
25 mL
Check Cart for Availability
€ 684,00
50 mL
Check Cart for Availability
€ 1.170,00

About This Item

NACRES:
NA.56
UNSPSC Code:
41106500

€ 314,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

manufacturer/tradename

Novagen®

storage condition

do not freeze

application(s)

life science and biopharma

shipped in

ambient

storage temp.

2-8°C

General description

GST•Bind Resin utilizes an 14-atom spacer arm to covalently attach reduced glutathione via a sulfide linkage. The high degree of substitution of glutathione ensures a high binding capacity with yields of glutathione S-transferase (GST) fusion proteins of 5 to 8 mg/ml settled resin. The resin can be re-used up to 6 times without loss of capacity.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.55013706918.55155
application(s)

life science and biopharma

application(s)

peptide synthesis

application(s)

life science and biopharma

application(s)

peptide synthesis

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

form

liquid

form

beads

form

liquid

form

powder

manufacturer/tradename

Novagen®

manufacturer/tradename

Novabiochem®

manufacturer/tradename

Novagen®

manufacturer/tradename

Novabiochem®

storage condition

do not freeze

storage condition

-

storage condition

do not freeze

storage condition

-

Quality Level

400

Quality Level

200

Quality Level

400

Quality Level

200


pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

flash_point_f

96.8 °F

flash_point_c

36 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

This article shows the use of BugBuster® and Benzonase® as protein purification tools to extract recombinant proteins from E. coli and to reduce the viscosity of the extract.

YeastBuster™ Protein Extraction Reagent for Fast, Efficient Extraction of Proteins from Yeast


Rahul Bhattacharjee et al.
Molecular biology of the cell, 31(9), 917-929 (2020-02-27)
In many organisms, positive and negative signals cooperate to position the division site for cytokinesis. In the rod-shaped fission yeast Schizosaccharomyces pombe, symmetric division is achieved through anillin/Mid1-dependent positive cues released from the central nucleus and negative signals from the



Global Trade Item Number

SKUGTIN
70541-404055977273663
70541-304055977273656
70541-504055977273670

Questions

Reviews

No rating value

Active Filters