Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 40 reactions | Check Cart for Availability | € 261,00 |
About This Item
NACRES:
NA.51
UNSPSC Code:
41106602
€ 261,00
Check Cart for Availability
form
liquid
Quality Segment
manufacturer/tradename
Novagen®
storage condition
OK to freeze
storage temp.
−20°C
General description
The pT7Blue™-2 T-Vector is designed for simplified cloning of PCR products. Many thermostable DNA polymerases, such as Taq DNA polymerase, leave single 3′ A-nucleotide overhangs on their reaction products. These molecules can be ligated directly to a vector containing compatible single T-nucleotide overhangs. The pT7Blue™-2 vector has been specifically constructed for thisapplication and has been prepared for T-cloning by digestion with EcoRV followed by the addition of single 3′ dT residues at each end. The T-Vector is provided ready to ligate with DNA amplified with any DNA polymerase that leaves single 3′ dA overhangs. Target inserts having other types of ends, such as reaction products of proofreading polymerase or restriction fragments, can be easily prepared for T-cloning with the Single dA<TMSYMBOL></TMSYMBOL> Tailing Kit.
The pT7Blue™-2 Vector is a versatile plasmid featuring the 43-bp Xenopus β-globin 5′-UTR for enhancement of translation in rabbit reticulocyte lysates and frog oocytes. Inserts cloned in-frame with the lacZ α-peptide coding sequence can also be expressed in E. coli from the lac promoter. The vector has many additional features that make it an excellent choice for a variety of applications.
The pT7Blue™-2 Vector is a versatile plasmid featuring the 43-bp Xenopus β-globin 5′-UTR for enhancement of translation in rabbit reticulocyte lysates and frog oocytes. Inserts cloned in-frame with the lacZ α-peptide coding sequence can also be expressed in E. coli from the lac promoter. The vector has many additional features that make it an excellent choice for a variety of applications.
Other Notes
- 2 g pT7Blue™-2 T-Vector, 50 ng/l
- 10 l Positive Control T-Vector Insert, 2.5 ng/l
Legal Information
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PT7BLUE is a trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
1 of 1
This Item | |||
|---|---|---|---|
| form liquid | form liquid | form liquid | form liquid |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −70°C |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| manufacturer/tradename Novagen® | manufacturer/tradename Novagen® | manufacturer/tradename Novagen® | manufacturer/tradename Novagen® |
| storage condition OK to freeze | storage condition OK to freeze | storage condition OK to freeze, avoid repeated freeze/thaw cycles | storage condition OK to freeze, avoid repeated freeze/thaw cycles |
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 69080-3 | 04055977259575 |



