Skip to Content
Merck

Skip To

6540-M

L-Tryptophan

Synonym(s):

L-Tryptophan, Trp

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 g
Check Cart for Availability
Inquire

About This Item

Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
UNSPSC Code:
12352209
NACRES:
NA.25
MDL number:
eCl@ss:
32160406

Inquire

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

RTECS - YN6130000
Merck USA index - 14, 9797

Quality Segment

assay

≥98.5% (titration)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 1 mg/mL

cation traces

heavy metals: ≤0.0015%

storage temp.

10-30°C

SMILES string

[nH]1c2c(c(c1)CC(N)C(=O)O)cccc2

InChI

1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)

InChI key

QIVBCDIJIAJPQS-UHFFFAOYSA-N

General description

An essential amino acid for human development. Precursor of serotonin.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1176T261000051145
form

crystalline solid

form

-

form

-

form

-

assay

≥98.5% (titration)

assay

-

assay

-

assay

-

storage temp.

10-30°C

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

-

solubility

water: 1 mg/mL

solubility

-

solubility

-

solubility

-

cation traces

heavy metals: ≤0.0015%

cation traces

-

cation traces

-

cation traces

-

color

white

color

-

color

-

color

white


Still not finding the right product?

Explore all of our products under L-Tryptophan


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
6540-100GM04055977184563

Questions

Reviews

No rating value

Active Filters