Skip to Content
Merck

Skip To

620-M

Giemsa Stain Solution Modified Azure Blend

HARLECO® Polychromatic stain used primarily with Wright Stain to stain blood cells and bone marrow specimens

Giemsa Stain Solution Modified Azure Blend

Synonym(s):

Hematology Stain

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41116121


Quality Level

form

liquid

transition temp

flash point 11 °C

λmax

520-530 nm (Eosin), 645-655 nm (Azure)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

Application

HARLECO® Polychromatic stain used primarily with Wright Stain to stain blood cells and bone marrow specimens
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Polychromatic stain used primarily with Wright Stain to stain blood cells and bone marrow specimens. Has various other uses in microbiology and histology staining.
Dilute with 50 volumes of distilled water or buffer immediately before use.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

51.8 °F

flash_point_c

11 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
620-7504061839405066
GF49946417-1EA04061825409559
GF49946417-2EA04061829500405
GF49946417-10EA04061829467241
GF49946417-5EA04061829560997

Questions

Reviews

Active Filters

  1. 1 Ratings-only review