Skip to Content
Merck
All Photos(1)

Key Documents

605204-M

Sigma-Aldrich

Thrombin, Immobilized, Human Plasma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

form

suspension

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

storage temp.

2-8°C

General description

Native thrombin from human plasma, bound to agarose beads. Thrombin is a key enzyme in the coagulation cascade. Converts fibrinogen to fibrin and activates factor XIII, which cross-links and stabilizes the fibrin polymer.

Warning

Toxicity: Standard Handling (A)

Physical form

In 100 mM NaCl, 20 mM Tris-HCl, ≤0.1% sodium azide, pH 7.4.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Other Notes

Xiang, B., et al. 1998. Proc. Natl. Acad. Sci. USA95, 7412.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service