Skip to Content
Merck
All Photos(1)

Key Documents

69671

Millipore

Thrombin, Restriction Grade

Synonym(s):

Thrombin, Restriction Grade

Sign Into View Organizational & Contract Pricing

Select a Size

50 U
€182.00

€182.00


Estimated to ship on26 May 2025



Select a Size

Change View
50 U
€182.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

€182.00


Estimated to ship on26 May 2025


biological source

human plasma

Quality Level

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

technique(s)

protein extraction: suitable

suitability

suitable for additive or modifier in the separation of proteins or peptides

application(s)

life science and biopharma

shipped in

wet ice

storage temp.

−20°C

General description

Restriction Grade Thrombin is a highly purified preparation qualified to specifically cleave target proteins produced with appropriate vectors. This preparation is functionally tested for activity with fusion proteins and is free of detectable contaminating proteases. Prepared from human plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV. Thrombin is supplied as a solution in 50% glycerol and includes 10X Thrombin Cleavage Buffer and a Cleavage Control Protein.

Components

•50 UThrombin

•1 ml10X Thrombin Cleavage Buffer

•2 ml1X Thrombin Dilution/Storage Buffer

•10 µgCleavage Control Protein

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Unit Definition

One unit is defined as the amount of enzyme needed to cleave 1 mg of fusion protein in 16 hours at 20°C in a 200 µl reaction containing 20 mM Tris-HCl pH 8.4, 150 mM NaCl, 2.5 mM CaCl₂, 50 µg fusion protein.

Preparation Note

Prepared from human plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jooyoung Park et al.
eLife, 8 (2019-12-20)
The computational design of a symmetric protein homo-oligomer that binds a symmetry-matched small molecule larger than a metal ion has not yet been achieved. We used de novo protein design to create a homo-trimeric protein that binds the C3 symmetric
Christian M Smolko et al.
Scientific reports, 9(1), 19409-19409 (2019-12-21)
Protein kinases are enzymes whose abundance, protein-protein interactions, and posttranslational modifications together determine net signaling activity in cells. Large-scale data on cellular kinase activity are limited, because existing assays are cumbersome, poorly sensitive, low throughput, and restricted to measuring one

Articles

Find highly-specific cleavage enzymes for isolating proteins during fusion protein purification including enterokinase, factor Xa, HRV 3C protease, and thrombin.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service