Skip to Content
Merck
All Photos(1)

Key Documents

5.31000

Sigma-Aldrich

Brk Inhibitor, Cpd 4f

Synonym(s):

Brk Inhibitor, Cpd 4f, PTK6 Inhibitor, Breast Tumor Kinase Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€205.00

€205.00


Estimated to ship on31 May 2025



Select a Size

Change View
10 MG
€205.00

About This Item

Empirical Formula (Hill Notation):
C17H13N3O
Molecular Weight:
275.30
UNSPSC Code:
12352200
NACRES:
NA.77

€205.00


Estimated to ship on31 May 2025


Assay

≥97% (HPLC)

Quality Level

form

powder

potency

3.15 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

General description

A cell permeable pyridoindole based compound that acts as a highly potent and selective inhibitor of breast tumor kinase (Brk; IC50 = 3.15 nM) with minimal or no effect against a panel of more than 30 other protein kinases (IC50 = <10 µM for PIM2, NEK1 and2, CAMK4, MAPKAPK3 and 5 and MARK1, 3, and 5). Does not affect the activity of Akt isoforms, AGC kinases, ERK1 and 2, JNK 1 and 2 and HIPK isoforms. Displays anti-proliferative activity against multiple breast cancer cell lines (GI50 = 990 nM; 1.02 µM, and 1.58 µM for MCF7, HS-578/T, BT-549, respectively).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Brk

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are good for up to 2 weeks at -70°C.
Use only fresh DMSO for reconstitution.

Other Notes

Mahmoud, K. A., et al. 2014. Bioorg. Med. Chem. Lett.24, 1948.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service