Skip to Content
Merck
All Photos(1)

Key Documents

09-279

Sigma-Aldrich

Anti-20S proteasome subunit β6 Antibody

serum, Upstate®

Synonym(s):

C5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€353.00

€353.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€353.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€353.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rabbit

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PSMB6(5694)

Specificity

Recognizes proteasome subunit β6

Immunogen

Peptide corresponding to residues 185-198 of human proteasome subunit β6 conjugated to KLH.

Application

Detect 20S proteasome subunit β6 using this Anti-20S proteasome subunit β6 Antibody validated for use in WB, IH.
Research Category
Protein Trafficking
Research Sub Category
Ubiquitin & Ubiquitin Metabolism

Target description

26 kDa

Physical form

100 µL of purified antiserum in PBS containing 0.01M sodium azide. Dilute to working strength with phosphate buffered saline pH 7.2-7.4.
Serum

Storage and Stability

Stable for 1 year at -20°C from date of shipment for up to 12 months.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service