Skip to Content
Merck
All Photos(1)

Key Documents

658550

Sigma-Aldrich

AG 1295

A cell-permeable, reversible, ATP-competitive, and selective inhibitor of platelet-derived growth factor (PDGF) receptor kinase (IC₅₀ = 500 nM) and PDGF-dependent DNA synthesis (IC₅₀ = 2.5 µM) in Swiss/3T3 cells.

Synonym(s):

AG 1295, 6,7-Dimethyl-2-phenylquinoxaline

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€139.00

€139.00


Estimated to ship on31 May 2025



Select a Size

Change View
5 MG
€139.00

About This Item

Empirical Formula (Hill Notation):
C16H14N2
CAS Number:
Molecular Weight:
234.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€139.00


Estimated to ship on31 May 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL
dichloromethane: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

n1c2c(ncc1c3ccccc3)cc(c(c2)C)C

InChI

1S/C16H14N2/c1-11-8-14-15(9-12(11)2)18-16(10-17-14)13-6-4-3-5-7-13/h3-10H,1-2H3

InChI key

FQNCLVJEQCJWSU-UHFFFAOYSA-N

General description

A cell-permeable, reversible, ATP-competitive, and selective inhibitor of platelet-derived growth factor (PDGF) receptor kinase (IC50 = 500 nM) and PDGF-dependent DNA synthesis (IC50 = 2.5 µM) in Swiss/3T3 cells. Has no effect on EGF receptor autophosphorylation and shows only a weak effect on EGF- or insulin-stimulated DNA synthesis. A potent blocker of smooth muscle cell proliferation.
Selectively inhibits platelet-derived growth factor (PDGF) receptor kinase (IC50 = 500 nM) and the PDGF-dependent DNA synthesis (IC50 = 2.5 µM) in Swiss 3T3 cells. Has no effect on epidermal growth factor (EGF) receptor autophosphorylation and shows only a weak effect on EGF- or insulin-stimulated DNA synthesis.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PDGFR kinase
Product competes with ATP.
Reversible: yes
Target IC50: 500 nM against PDGFR kinase

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Banai, S., et al. 1998. Circulation 97, 1960.
Levitzki, A., and Gazit, A. 1995. Science267, 1782.
Kovalenko, M., et al. 1994. Cancer Res.54, 6106.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service