Skip to Content
Merck

Skip To

528906

Polyinosinic Acid • Polycytidylic Acid, Sodium Salt, Double-Stranded

Known to induce interferon production. Hypochromicity: ≥40% at 248 nm. Extinction coefficient = 5000 M-1cm-1 at 260 nm, pH 7.0.

Synonym(s):

Polyinosinic Acid • Polycytidylic Acid, Sodium Salt, Double-Stranded, Poly I • poly C, Na

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
Inquire

About This Item

CAS Number:
UNSPSC Code:
12352208
NACRES:
NA.51
MDL number:

Inquire

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

RTECS - TR0175000

assay

≥99% (TLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

aqueous buffer: 10 mg/mL, water: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na].[P](=O)(OC[C@H]3O[C@H]([C@@H]([C@@H]3O)O)[n]4c5ncnc(c5nc4)O)(O)O.[P](=O)(OC[C@H]1O[C@H]([C@@H]([C@@H]1O)O)N2C=CC(=NC2=O)N)(O)O

InChI

1S/C10H13N4O8P.C9H14N3O8P.Na/c15-6-4(1-21-23(18,19)20)22-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18;/h2-4,6-7,10,15-16H,1H2,(H,11,12,17)(H2,18,19,20);1-2,4,6-8,13-14H,3H2,(H2,10,11,15)(H2,16,17,18);/t

InChI key

GETQKWBYQAUFQM-VRQAYDGLSA-N

General description

Hypochromicity: ≥40% at 248 nm.
Known to induce interferon production.

Application


  • Penaeus monodon Interferon Regulatory Factor (PmIRF) Activates IFNs and Antimicrobial Peptide Expression via a STING-Dependent DNA Sensing Pathway.: This study explores the role of PmIRF in shrimp immune responses, specifically how it activates interferons and antimicrobial peptides through the STING-dependent DNA sensing pathway, enhancing antiviral defenses. (Soponpong et al., 2021).

  • Identification, expression profiling of a grass carp TLR8 and its inhibition leading to the resistance to reovirus in CIK cells.: This article focuses on the TLR8 receptor in grass carp, its expression profiles, and how its inhibition confers resistance to reovirus in fish cell lines, providing insights into antiviral immunity in aquatic species. (Chen et al., 2013).

  • Identification and expression profiles of ADAR1 gene, responsible for RNA editing, in responses to dsRNA and GCRV challenge in grass carp (Ctenopharyngodon idella).: The study examines the ADAR1 gene involved in RNA editing in grass carp, highlighting its response to double-stranded RNA and grass carp reovirus, shedding light on fish immune mechanisms. (Yang et al., 2012).

  • Immune activation and target organ damage are consequences of hydrodynamic treatment but not delivery of naked siRNAs in mice.: This research differentiates the effects of hydrodynamic treatment and naked siRNA delivery in mice, showing that immune activation and organ damage are associated with the treatment method rather than the siRNAs themselves. (Rácz et al., 2011).

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P0913P1530P4929
assay

≥99% (TLC)

assay

(Free nucleotides NMT 1% (TLC)), ≥99% (less than 1% free nucleotides, TLC)

assay

(Free nucleotides NMT 1% (TLC)), ≥99% (less than 1% free nucleotides, TLC)

assay

-

Quality Level

100

Quality Level

300

Quality Level

300

Quality Level

200

form

powder

form

lyophilized powder

form

lyophilized powder

form

powder

solubility

aqueous buffer: 10 mg/mL, water: soluble

solubility

H2O: 10 mg/mL

solubility

H2O: 10 mg/mL

solubility

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage condition

OK to freeze

storage condition

desiccated

storage condition

desiccated

storage condition

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters