Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 mg | Check Cart for Availability | € 497,00 |
About This Item
NACRES:
NA.77
UNSPSC Code:
12352202
description
Merck USA index - 14, 7521
Quality Segment
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, avoid repeated freeze/thaw cycles
foreign activity
Plasmin, none detected (chromogenic substrate S-2251)
shipped in
wet ice
storage temp.
−70°C
General description
Native Lys-type plasminogen from human plasma prepared from homogeneous Glu-plasminogen by activation with plasmin. Activation results in the release of a 76 residue peptide (Glu1 - Lys76) fragment. Lys77-plasminogen can be readily converted to Lys77-plasmin by any of the common plasminogen activators.
Physical form
In 100 mM NaCl, 50 mM Tris-HCl, pH 7.4.
Preparation Note
Following initial thaw, aliquot and freeze (-70°C).
Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
Analysis Note
Homogeneous by SDS-PAGE
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Toxicity: Irritant (B)
1 of 1
This Item | |||
|---|---|---|---|
| form liquid | form lyophilized | form liquid | form lyophilized |
| shipped in wet ice | shipped in ambient | shipped in wet ice | shipped in ambient |
| storage condition OK to freeze, avoid repeated freeze/thaw cycles | storage condition OK to freeze | storage condition OK to freeze, avoid repeated freeze/thaw cycles | storage condition OK to freeze |
| storage temp. −70°C | storage temp. −20°C | storage temp. −70°C | storage temp. −20°C |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® |
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



