Skip to Content
Merck

Skip To

5.08317

A740003

Synonym(s):

A740003, N-(1-{[(cyanoimino)(5-quinolinylamino) methyl] amino}-2,2-dimethylpropyl)-2-(3,4-dimethoxyphenyl)acetamide, A 740003, P2X7 Purinegic Receptor, A740003, A740003

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C26H30N6O3
CAS Number:
Molecular Weight:
474.55
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Quality level:
Storage condition:
OK to freeze, protect from light

assay

≥98% (HPLC)

Quality Level

form

powder

potency

18 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

DMSO: 30 mg/mL

storage temp.

2-8°C

InChI

1S/C26H30N6O3/c1-26(2,3)24(31-23(33)15-17-11-12-21(34-4)22(14-17)35-5)32-25(29-16-27)30-20-10-6-9-19-18(20)8-7-13-28-19/h6-14,24H,15H2,1-5H3,(H,31,33)(H2,29,30,32)

Looking for similar products? Visit Product Comparison Guide

General description

A highly potent, selective, competitive antagonist of P2X7 purinegic receptor (IC50 = 18 nM and 40 nM, respectively as measured by Bz-ATP-stimulated Ca2+ flux). Does not affect other P2 receptors even at 10 µM concentrations. Blocks agonist-evoked IL-1b release (IC50 = 156 nM) and pore formation (IC50 = 92 nM) in differentiated human THP-1 cells. Also shown to reduce carrageenan and Freund′s adjuvant -induced thermal hyperalgesia (ED50 = 38-54 mg/kg, i.p) and neuropathic pain induced by chronic constriction injury (CIC) of the sciatic nerve.

Biochem/physiol Actions

Primary Target
P2X7

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Slight warming is required for complete solubilization.

Other Notes

Honore, P., et al. 2006. J. Pharmacol. Exp. Ther.319, 1376.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
508317000104055977261851

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service