Skip to Content
Merck
All Photos(1)

Key Documents

499260

Sigma-Aldrich

Osteopontin, His•Tag®, Mouse, Recombinant, Mouse

Synonym(s):

rmOPN

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€565.00

€565.00


Estimated to ship on21 May 2025



Select a Size

Change View
50 μG
€565.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.45

€565.00


Estimated to ship on21 May 2025


Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤100 pg/μg Endotoxins (pg/μg rmOPN)

shipped in

ambient

storage temp.

−70°C

General description

M.W. 65,000 and 30,000 under reducing conditions.
Recombinant, mouse osteopontin fused at the C-terminus to a HisTag sequence and expressed in NSO-derived Leu17-Asn294 (Glu99Gly) murine myeloma cells. A secreted, highly acidic, calcium-binding, RGD-containing, phosphorylated glycoprotein. Osteopontin is expressed in several cell types including macrophages, fibroblasts, epithelial cells, T cells and osteoclasts in response to activation by cytokines, growth factors or inflammatory mediators. Mouse OPN cDNA encodes a 294 amino acid protein that has a 16 amino acid predicted signal peptide, which is cleaved to generate 278 amino acid mature protein. The mature protein contains an integrin binding arginine-glycine-aspartic acid (RGD) sequence. Exhibits some renoprotective actions in renal injury, such as increasing tolerance to acute ischemia, inhibiting inducible nitric oxide synthase, and reducing cell peroxide levels.

Biochem/physiol Actions

~200 ng immobilized osteopontin/well will mediate the adhesion of ≥25% of the 293 cells plated at 10⁵ cells/well

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm sterile-filtered PBS, 50 µg BSA/µg rmOPN.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 1 month at 4°C and for up to 3 months at -70°C. Avoid freeze/thaw cycles of stock solutions.
Reconstitute to ≥100 µg/ml with sterile PBS.

Other Notes

Xie, Y., et al. 2001. Kidney Int.60, 1645.
Chiba, S., et al. 2000. Microbiol. Immunol. 44, 319.
Miyazaki, Y., et al. 1990. J. Biol. Chem. 265, 14432.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristen Johnson et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 18(6), 994-1004 (2003-06-24)
Osteopontin and PP(i) both suppress hydroxyapatite deposition. Extracellular PP(i) deficiency causes spontaneous hypercalcification, yet unchallenged osteopontin knockout mice have only subtle mineralization abnormalities. We report that extracellular PP(i) deficiency promotes osteopontin deficiency and correction of osteopontin deficiency prevents hypercalcification, suggesting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service