Skip to Content
Merck

Skip To

401484

IKK-2 Inhibitor IV

InSolution, ≥95%

Synonym(s):

InSolution IKK-2 Inhibitor IV

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 μg
Check Cart for Availability
€ 139,00

About This Item

Empirical Formula (Hill Notation):
C12H10FN3O2S
Molecular Weight:
279.29
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
liquid
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light

€ 139,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

liquid

potency

18 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

color

clear pale yellow

shipped in

wet ice

storage temp.

2-8°C

General description

IKK-2 Inhibitor IV (Cat. No. 401481) supplied in DMSO. A cell-permeable and potent inhibitor of IKK-2 (IC50 = 18 nM) that exhibits selectivity over IKK-1, JNK, and p38 MAPK. Inhibits TNF-α production in human monocytes (IC50 in the range of 0.15-2.5 µM) and blocks IL-8 and IL-6 production by synovial fibroblasts (IC50 = 100 nM). Shown to reduce paw oedema in rat arthritis model (~100% inhibition at a dose of 30 mg/kg).

Biochem/physiol Actions

Cell permeable: yes
Product competes with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Physical form

A 10 mM (500 µg/179 µl) solution of IKK-2 Inhibitor IV (Cat. No. 401481) in DMSO.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
401481401485124015
form

liquid

form

solid

form

liquid

form

liquid

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage condition

OK to freeze, protect from light, desiccated (hygroscopic)

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

storage condition

OK to freeze, desiccated (hygroscopic), protect from light


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

188.6 °F - closed cup - (Dimethylsulfoxide)

flash_point_c

87 °C - closed cup - (Dimethylsulfoxide)



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters