Skip to Content
Merck

Skip To

488285

n-Nonyl-β-D-glucopyranoside

Mild detergent especially suitable for isolation of membrane-bound proteins with minimal denaturation.

Synonym(s):

n-Nonyl-β-D-glucopyranoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H30O6
CAS Number:
Molecular Weight:
306.40
NACRES:
NA.25
UNSPSC Code:
12352200
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated

color

white

CMC

6.5 mM

solubility

water: 250 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C15H30O6/c1-2-3-4-5-6-7-8-9-20-15-14(19)13(18)12(17)11(10-16)21-15/h11-19H,2-10H2,1H3/t11-,12-,13+,14-,15-/m1/s1

InChI key

QFAPUKLCALRPLH-UXXRCYHCSA-N

General description

Mild detergent especially suitable for isolation of membrane-bound proteins with minimal denaturation (CMC: 6.5 mM).

Other Notes

Chung, Y.J., and Jeffrey, G.A. 1989. Biochim. Biophys. Acta985, 300.
de Grip, W.J., and Bovee-Geurts, P.H.M. 1979. Chem. Phys. Lipids23, 321.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
488285-1GM04055977200485

Questions

Reviews

No rating value

Active Filters