Skip to Content
Merck
All Photos(2)

Key Documents

480747

Sigma-Aldrich

Neurogenesis Inducer V, KHS101

A blood brain barrier permeable pyrimidinyl-aminothiazole compound that specifically interacts with TACC3 (transforming acidic coiledcoil-containing protein 3) and promotes nuclear localization of ARNT2 (aryl-hydrocarbon receptor nuclear translocator 2).

Synonym(s):

Neurogenesis Inducer V, KHS101, N⁴-Isobutyl-N²-((2-phenylthiazol-4-yl)methyl)pyrimidine-2,4-diamine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€126.00

€126.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€126.00

About This Item

Empirical Formula (Hill Notation):
C18H21N5S
CAS Number:
Molecular Weight:
339.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€126.00


Estimated to ship on17 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

resin (Sticky)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

2-8°C

SMILES string

[s]1c(nc(c1)CNc3nc(ccn3)NCC(C)C)c2ccccc2

InChI

1S/C18H21N5S/c1-13(2)10-20-16-8-9-19-18(23-16)21-11-15-12-24-17(22-15)14-6-4-3-5-7-14/h3-9,12-13H,10-11H2,1-2H3,(H2,19,20,21,23)

InChI key

DGRJOOOHPBSAHD-UHFFFAOYSA-N

General description

A blood brain barrier permeable pyrimidinyl-aminothiazole compound that specifically interacts with TACC3 (transforming acidic coiledcoil-containing protein 3) and promotes nuclear localization of ARNT2 (aryl-hydrocarbon receptor nuclear translocator 2). Shown to significantly induce neuronal differentiation of cultured rat hippocampal NPCs (neural progenitor cells; EC50 ~ 1 µM) and in adult rats (6 mg/kg, s.c.) and suppress proliferation.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Wurdak, H., et al. 2010. Proc. Natl. Acad. Sci. USA107, 16542

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service