Z678740
FORTUNA® Polyfix® bottle top dispenser with PTFE coated piston
dispensing volume 2-10 mL
Select a Size
1 EA
€236.00
€236.00
Check Cart for Availability
All Photos(1)
Select a Size
Change View
1 EA
€236.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
€236.00
Check Cart for Availability
manufacturer/tradename
Fortuna Graf 101.071-38
dispensing volume measuring range
2-10 mL
volume
2-10 mL
fitting
GL 40
GL 45
General description
For dispensing of weak acids and alkaline solutions
Features:
Features:
- Visible dispensing process by the use of a dispensing cylinder made of glass, coated with PP-protection against breakage
- Coated PTFE-piston
- Ergonomic finger protection cap
- Fast and easy volume setting with additional fine adjustment
- FIX adapter made of PP
- Valve block made of PP, no metal springs in the valve - no corrosion
Legal Information
FORTUNA is a registered trademark of Poulten & Graf Ltd.
POLYFIX is a registered trademark of Poulten & Graf Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)