Skip to Content
Merck
All Photos(3)

Key Documents

AB5539

Sigma-Aldrich

Anti-Neurofilament H (200 kDa) Antibody

CHEMICON®, chicken polyclonal

Synonym(s):

NF-H

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€651.00

€651.00


Estimated to ship on02 June 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1347

Request a Bulk Order

Select a Size

Change View
50 μL
€651.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€651.00


Estimated to ship on02 June 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1347

Request a Bulk Order

Product Name

Anti-Neurofilament H Antibody, Chemicon®, from chicken

biological source

chicken

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, pig, bovine, feline, human, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

bovine ... Nefh(528842)
cat ... Nefh(101087272)
human ... NEFH(4744)
mouse ... Nefh(380684)
pig ... Nefh(100156492)
rat ... Nefh(24587)

Specificity

Recognizes Neurofilament-Heavy (NF-H). The antibody stains sharply defined cytoplasmic filaments in neuronal cell bodies and especially axons.

Immunogen

Purified bovine NF-H.

Application

Anti-Neurofilament H Antibody is an antibody against Neurofilament for use in IC, IH, IH(P) & WB.
Western blot: 1:1,000-1:10,000 using ECL. Recognizes a band of 200-220 kDa.

Immunohistochemistry: 1:1,000-1:10,000 on frozen or paraffin embedded tissue sections. Suggested fixative is 4% paraformaldehyde.

Immunocytochemistry: 1:1,000-1:10,000 on cells in tissue culture fixed for one minute in 3.7% formalin and one minute in -20°C methanol.

Optimal working dilutions must be determined by the end user.

Target description

200 kDa

Physical form

Format: Purified
Purified chicken IgY in PBS containing 5mM sodium azide.

Analysis Note

Control
Positive control: brain.

Negative control: kidney, liver.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Computation identifies structural features that govern neuronal firing properties in slowly adapting touch receptors.
Lesniak, DR; Marshall, KL; Wellnitz, SA; Jenkins, BA; Baba, Y; Rasband, MN; Gerling, GJ; Lumpkin, EA
eLife null
Juan Miguel Jimenez-Andrade et al.
Bone, 46(2), 306-313 (2009-09-22)
Although musculoskeletal pain is one of the most common causes of chronic pain and physical disability in both developing and developed countries, relatively little is known about the nerve fibers and mechanisms that drive skeletal pain. Small diameter sensory nerve
Zu-Lin Chen et al.
Developmental biology, 327(1), 158-168 (2009-01-03)
Laminins have dramatic and varied actions on neurons in vitro. However, their in vivo function in brain development is not clear. Here we show that knockout of laminin gamma1 in the cerebral cortex leads to defects in neuritogenesis and neuronal
Lucas M Viana et al.
Hearing research, 327, 78-88 (2015-05-24)
Recent animal work has suggested that cochlear synapses are more vulnerable than hair cells in both noise-induced and age-related hearing loss. This synaptopathy is invisible in conventional histopathological analysis, because cochlear nerve cell bodies in the spiral ganglion survive for
Katarzyna Dover et al.
Nature communications, 7, 12895-12895 (2016-09-27)
Neurons in vertebrate central nervous systems initiate and conduct sodium action potentials in distinct subcellular compartments that differ architecturally and electrically. Here, we report several unanticipated passive and active properties of the cerebellar granule cell's unmyelinated axon. Whereas spike initiation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service