Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C13H9ClN2O6S
CAS Number:
Molecular Weight:
356.74
UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Pricing and availability is not currently available.
Quality Level
assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
solubility
DMSO: 50 mg/mL
shipped in
ambient
storage temp.
−20°C
SMILES string
[S](=O)(=O)(Nc2cc(ccc2)[N+](=O)[O-])c1c(ccc(c1)C(=O)O)Cl
InChI
1S/C13H9ClN2O6S/c14-11-5-4-8(13(17)18)6-12(11)23(21,22)15-9-2-1-3-10(7-9)16(19)20/h1-7,15H,(H,17,18)
InChI key
IIJQJWNGBILZCU-UHFFFAOYSA-N
General description
792949 is the first purely competitive inhibitor of mitochondrial Citrate Transport Protein (CTP), it produced 85% inhibition of citrate transport at 1mM.
A competitive inhibitor of mitochondrial citrate transport protein (CTP). Exhibits 85% inhibition at 1 mM.
Preparation Note
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Aluvila, S., et al. 2010. Mol. Pharmacol.77, 26.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Storage Class
11 - Combustible Solids
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 475877-5MG | 04055977203455 |