Skip to Content
Merck
All Photos(1)

Key Documents

428010

Millipore

Lauroylsarcosine, Sodium Salt

Ionic detergent used to purify membrane proteins. Also decreases transcription efficiency. Aggregation no.: 104.

Synonym(s):

Lauroylsarcosine, Sodium Salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
Inquire


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
Inquire

About This Item

Empirical Formula (Hill Notation):
C15H28NO3 · Na
CAS Number:
Molecular Weight:
293.38
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.21


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

description

Merck USA index - 14, 4368

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 1 M

foreign activity

DNAse, RNAse, Protease, Phosphatase, none detected

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Na+].N(CC(=O)[O-])(C)C(=O)CCCCCCCCCCC

InChI

1S/C15H29NO3.Na/c1-3-4-5-6-7-8-9-10-11-12-14(17)16(2)13-15(18)19;/h3-13H2,1-2H3,(H,18,19);/q;+1/p-1

InChI key

KSAVQLQVUXSOCR-UHFFFAOYSA-M

General description

Ionic detergent used to purify membrane proteins. Also decreases transcription efficiency. Aggregation no.: 104

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store at room temperature (20°C). Stock solutions are stable to up to 6 months at 20°C.

Other Notes

Rao, J.N., et al. 2009. J. Mol. Biol.390, 516.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

512.6 °F - closed cup

Flash Point(C)

267 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service