Skip to Content
Merck
All Photos(1)

Documents

8.14715

Sigma-Aldrich

N-Lauryl sarcosine sodium salt

for synthesis

Synonym(s):

N-Lauryl sarcosine sodium salt, Sarcosyl, Sodium lauroylsarcosinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3-(CH2)10-CO-N(CH3)-CH2-COONa
CAS Number:
Molecular Weight:
293.38
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-281-5
NACRES:
NA.22

vapor pressure

0.02 hPa ( 20 °C)

Quality Level

form

powder

potency

2888 mg/kg LD50, oral (Rat)

pH

8 (20 °C, 30 g/L in H2O)

mp

146 °C

transition temp

flash point 267 °C

density

1.14 g/cm3 at 20 °C

bulk density

400 kg/m3

storage temp.

2-30°C

InChI

1S/C15H29NO3.Na/c1-3-4-5-6-7-8-9-10-11-12-14(17)16(2)13-15(18)19;/h3-13H2,1-2H3,(H,18,19);/q;+1/p-1

InChI key

KSAVQLQVUXSOCR-UHFFFAOYSA-M

Related Categories

Application


  • Polystyrene adsorbents: This study highlights the use of N-lauryl sarcosine sodium salt for the extraction of functionally active recombinant proteins from inclusion bodies (SK Palanirajan, P Govindasamy, SN Gummadi - Scientific Reports, 2020).

  • Modified magnetite nanoparticles for selective trace determination: Discusses the preparation of N-Lauryl sarcosine sodium salt modified magnetite nanoparticles as an extractant for selective trace determination (H Eskandari, H Ashrafi - 2014).

  • On-plate spot test for identification and separation of surfactants: Mentions the use of N-lauryl sarcosine sodium salt in the formulation of spot tests for various surfactants (A Mohammad, A Siddiq, R Mobin, E Gaber - Journal of Industrial and …, 2014).

  • Influence of HSA on micellization: This paper explores the influence of human serum albumin on the micellization properties of N-lauryl sarcosine sodium salt and other surfactants (I Fatma, V Sharma, NA Malik, H Assad… - Journal of Molecular …, 2022).

  • Ultrasonic synthesis of stable oil filled microcapsules: This study explores the ultrasonic synthesis of oil-filled microcapsules using thiolated chitosan, potentially involving derivatives like ᴅʟ-N-Acetylhomocysteine thiolactone for improved stability and functional properties. The research provides insights into advanced microencapsulation techniques relevant in pharmaceutical and industrial applications (Link to the study).

Analysis Note

Assay (HClO₄): ≥ 95.0 %
Water (K. F.): ≤ 5.0 %
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

512.6 °F - closed cup

Flash Point(C)

267 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service