Skip to Content
Merck

Skip To

4100-OP

Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid

OmniPur® Grade, >=97.0%

Synonym(s):

OmniPur® EGTA, Ethylene Glycol bis (2-aminoethyl ether)-N,N,N′,N′-tetraacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
50 g
Check Cart for Availability
€ 197,00

About This Item

CAS Number:
UNSPSC Code:
12352106
NACRES:
NA.21
MDL number:

€ 197,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

crystalline powder (Fine)

reaction suitability

reagent type: chelator

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

foreign activity

DNAse, none detected, Protease, none detected, RNAse, none detected

shipped in

ambient

SMILES string

N(CCOCCOCCN(CC(=O)O)CC(=O)O)(CC(=O)O)CC(=O)O

InChI

1S/C14H24N2O10/c17-11(18)7-15(8-12(19)20)1-3-25-5-6-26-4-2-16(9-13(21)22)10-14(23)24/h1-10H2,(H,17,18)(H,19,20)(H,21,22)(H,23,24)

InChI key

DEFVIWRASFVYLL-UHFFFAOYSA-N

General description

Molecular biology grade.

Specifications:
Assay: ≥97.0%
DNase: None detected
Melting point (with decomposition): 238°C min
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E3889E4378E0396
Quality Level

100

Quality Level

400

Quality Level

400

Quality Level

400

form

crystalline powder (Fine)

form

powder

form

powder

form

powder

foreign activity

DNAse, none detected, RNAse, none detected, Protease, none detected

foreign activity

DNAse (exonuclease), NICKase (endonuclease), RNAse, and protease, none detected

foreign activity

-

foreign activity

-

reaction suitability

reagent type: chelator

reaction suitability

reagent type: chelator

reaction suitability

reagent type: chelator

reaction suitability

reagent type: chelator

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

storage condition

OK to freeze

storage condition

-

storage condition

-

storage condition

-


Storage Class

11 - Combustible Solids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
4100-50GM04055977242027

Questions

Reviews

No rating value

Active Filters