Skip to Content
Merck

Skip To

401477

IκB Kinase Inhibitor Peptide, Cell-Permeable

The IκB Kinase Inhibitor Peptide, Cell-Permeable controls the biological activity of IκB Kinase. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Synonym(s):

IκB Kinase Inhibitor Peptide, Cell-Permeable, Ac- AAVALLPAVLLALLAPDDRHDSGLDSMKDE-NH2, Ac-AAVALLPAVLLALLAPDDRHDSGLDSMKDE-NH2

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C138H229N37O45S
Molecular Weight:
3158.58
NACRES:
NA.77
UNSPSC Code:
12352202
Assay:
≥95% (HPLC)
Form:
lyophilized
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light


Quality Segment

assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

solubility

PBS, pH 7.4: 1 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A 14-amino acid peptide corresponding to the active IκB phosphorylation recognition sequence fused to the hydrophobic region of the fibroblast growth factor signal peptide to aid in cellular delivery. Specifically inhibits LPS-induced IκB degradation by IκB kinases (IKK) in RAW 264.7 cells (<50 µg/ml) and prevents NF-κB activation. Supplied as a trifluoroacetate salt.

Biochem/physiol Actions

Cell permeable: no
Primary Target
LPS-induced IκB degradation by IKK in RAW 264.7 cells
Product does not compete with ATP.
Reversible: no
Target IC50: <50 µg/ml for inhibiting LPS-induced IκB degradation by IκB kinases (IKK) in RAW 264.7 cells and prevents NF-κB activation

Packaging

Packaged under inert gas

Physical form

Supplied as a trifluoroacetate salt.

Preparation Note

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ac-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Asp-Asp-Arg-His-Asp-Ser-Gly-Leu-Asp-Ser-Met-Lys-Asp-Glu-NH₂
Swaroop, N., et al. 2001. Pharm. Res.18, 1631.
Traenckner, E.B., et al. 1995. EMBO. J.14, 2876.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
401478613570328000
assay

≥95% (HPLC)

assay

≥97% (HPLC)

assay

≥97% (HPLC)

assay

≥95% (HPLC)

form

lyophilized

form

solid

form

lyophilized solid

form

lyophilized solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

solubility

PBS, pH 7.4: 1 mg/mL

solubility

water: 1 mg/mL

solubility

DMSO: 5 mg/mL

solubility

water: 1 mg/mL, DMSO: 5 mg/mL


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
401477-1MG04055977211924

Questions

Reviews

No rating value

Active Filters