Skip to Content
Merck

Skip To

4010CBC

Hypoxanthine

Xanthine oxidase substrate.

Synonym(s):

Hypoxanthine, 6-Hydroxypurine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 g
Check Cart for Availability
Inquire

About This Item

Empirical Formula (Hill Notation):
C5H4N4O
CAS Number:
Molecular Weight:
136.11
UNSPSC Code:
12352208
NACRES:
NA.51
MDL number:

Inquire

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

1 M HCl: 20 mg/mL, clear (faint color solution), DMF: 5 mg/mL, cloudy (with particulates), DMSO: 5 mg/mL, clear, colorless (with few particulates), hydrogen: 5 mg/mL, cloudy (with particulates)

cation traces

heavy metals: ≤10 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

[nH]1c2nc[nH][c](c2nc1)=O

InChI

1S/C5H4N4O/c10-5-3-4(7-1-6-3)8-2-9-5/h1-2H,(H2,6,7,8,9,10)

InChI key

FDGQSTZJBFJUBT-UHFFFAOYSA-N

General description

Please note: this product is the same product that was previously found using Cat. No. 4010. Only the base catalog number has been changed.
Xanthine oxidase substrate. A product of nucleic acid breakdown. Acts to salvage free purines for nucleic acid and nucleotide recycling. Used in conjunction with thymidine and aminopterin for cloning and selection of hybridoma cell lines.
Xanthine oxidase substrate. Formed in the animal body during nucleic acid breakdown. Acts to salvage free purines for nucleic acid and nucleotide recycling. Used in conjunction with thymidine and aminopterin for cloning and selection of hybridoma cell lines.

Application


  • Targeted metabolomics analysis approach to unravel the biofilm formation pathways of Enterococcus faecalis clinical isolates.: This research employs targeted metabolomics to understand the role of hypoxanthine in the biofilm formation pathways of Enterococcus faecalis, offering insights that could guide the development of new antibacterial strategies (Suriyanarayanan T et al., 2024).

Preparation Note

Following reconstitution, sterilize by filtration through a 0.22 µm filter, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
H93771191226Y0000441
Quality Level

100

Quality Level

200, 300

Quality Level

-

Quality Level

-

assay

≥99% (HPLC)

assay

≥99.0%

assay

-

assay

-

form

solid

form

powder

form

-

form

-

storage temp.

15-25°C

storage temp.

-

storage temp.

-

storage temp.

2-8°C

solubility

1 M HCl: 20 mg/mL, clear (faint color solution), DMSO: 5 mg/mL, clear, colorless (with few particulates), DMF: 5 mg/mL, cloudy (with particulates), hydrogen: 5 mg/mL, cloudy (with particulates)

solubility

-

solubility

-

solubility

-

storage condition

OK to freeze

storage condition

-

storage condition

-

storage condition

-


Still not finding the right product?

Explore all of our products under Hypoxanthine


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
4010CBC-25GM04055977189353

Questions

Reviews

No rating value

Active Filters