Skip to Content
Merck

Skip To

35-002

STK Antibody, 20X

Upstate®, from mouse

Synonym(s):

Anti-MPPH, Anti-MPPH2, Anti-PKB-GAMMA, Anti-PKBG, Anti-PRKBG, Anti-RAC-PK-gamma, Anti-RAC-gamma, Anti-STK-2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
237 μL
Check Cart for Availability
€ 583,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
monoclonal
Application:
IP
Citations:
-

€ 583,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

monoclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... AKT3(10000)

Immunogen

a proprietary formulation containing STK antibody at 20X concentration for use in KinEASE fluorescence polarization assays

Application

STK Antibody, 20X detects level of STK & has been published & validated for use in IP.

Analysis Note

routinely used as an STK antibody in KinEASE fluorescence polarization assays

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
05-591SAB2108761AMAB90835
clone

monoclonal

clone

SKB1, monoclonal

clone

polyclonal

clone

CL0899, monoclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

affinity purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

Gene Information

human ... AKT3(10000)

Gene Information

mouse ... Akt1(11651)

Gene Information

human ... SLC25A4(291)

Gene Information

human ... AKT1(207)

species reactivity

human

species reactivity

rat, human, mouse

species reactivity

dog, guinea pig, bovine, mouse, human, rabbit, rat, sheep, goat

species reactivity

human

biological source

mouse

biological source

mouse

biological source

rabbit

biological source

mouse


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
35-00204053252408861

Questions

Reviews

No rating value

Active Filters