Skip to Content
Merck

Skip To

252000

DDMAB

Water-soluble zwitterionic detergent with properties similar to DDMAU but differing in the length of the polymethylene arm.

Synonym(s):

DDMAB, N-Dodecyl-N,N-(dimethylammonio)butyrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
5 g
Check Cart for Availability
Inquire

About This Item

Empirical Formula (Hill Notation):
C18H37NO2
CAS Number:
Molecular Weight:
299.49
UNSPSC Code:
12161902
NACRES:
NA.21
MDL number:

Inquire

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥98% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic)

color

white to beige

CMC

4.3 mM

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

[N+](CCCCCCCCCCCC)(CCCC(=O)[O-])(C)C

InChI

1S/C18H37NO2/c1-4-5-6-7-8-9-10-11-12-13-16-19(2,3)17-14-15-18(20)21/h4-17H2,1-3H3

InChI key

ZFVNBVQBIZJUEY-UHFFFAOYSA-N

General description

Water-soluble zwitterionic detergent with properties similar to DDMAU (Cat. No. 252005 ) but differing in the length of the polymethylene arm. More efficient for the extraction of mycoplasma membrane antigens under mild conditions than DDMAU.

Preparation Note

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 6 weeks at 4°C.

Other Notes

Brenner, C., et al. 1995. Anal. Biochem. 224, 515.
Chevalier, Y., et al. 1991. Langmuir7, 848.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
00413D2004382310
form

crystalline solid

form

-

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥94%

assay

≥99% (TLC)

assay

98%

storage temp.

15-25°C

storage temp.

-

storage temp.

room temp

storage temp.

-

solubility

water: 1 mg/mL

solubility

-

solubility

ethanol: 50 mg/mL

solubility

-

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

200

CMC

4.3 mM

CMC

4.3

CMC

-

CMC

-


Still not finding the right product?

Explore all of our products under DDMAB


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
252000-5GM04055977217285

Questions

Reviews

No rating value

Active Filters